Print…
Luvas resistentes ao corte, HyFlex® 72-285 / 72-286
Fornecedor: Ansell
72-285/7 72-285/10 72-285/6 72-285/8 72-285/9 72-286/10 72-286/6 72-286/7 72-286/8 72-286/9
112-0234EA 141 EUR
112-0234 112-0232 112-0233 112-0235 112-0236 112-0237 112-0238 112-0239 112-0240 112-0241
Luvas resistentes ao corte, HyFlex® 72-285 / 72-286
Luvas
These gloves are knitted from a combination of Polyester, spandex and Dyneema® fibres.
  • Excellent cut resistance
  • Machine washable up to 90 °C
  • Long lasting with improved wear resistance
  • Improved heat insulation for greater comfort in cold work areas
Suitable for applications involving direct contact with food, e.g. boning, carving, fish processing, meals preparation, as well as fruit and vegetable processing.
Colour: Light blue
EN 388, Cat. II
Food contact approved
Certificações: 72-285: EN 388:2016 (1X4XB); Cat. II
72-286: EN 388:2016 (2X4XD);Mais informação do produto
Encomendar
ESPECIFICAÇÕES
Modelo 72-285 72-286
Ambidestras/Específicas Ambidestras Ambidestras
Environmentally Preferable
Descartável/Reutilizável Reutilizável Reutilizável
Medidor 13 10
Estilo do punho Tricotado Tricotado
Norma EN EN 388:2016 (1X4XB) EN 388:2016 (2X4XD)
Aprovadas para contacto com alimentos + +