Print…
Funil para juntas cónicas esmeriladas
Fornecedor: Brand
94910 94905 94915
241-0232EA 78 EUR
241-0232 241-0231 BRAA94915
Funil para juntas cónicas esmeriladas
Funis Funis de filtração Funis para juntas cónicas
De PP, translúcidos.
  • Ideal for transferring liquids or powders to multiple-neck flasks
  • Excellent chemical resistance
  • Excellent temperature resistance
  • Autoclavable
The funnel body is flattened on the side and the end of the shaft is shaped to fit conical ground joints.