Print…
Funnels, industrial, Azlon®
Fornecedor: SciLabware
FWC102 FWC106 FWC110 FWC116 FWC108 FWC104 FWC112 FWC118 FWC114
221-0037EA 38 EUR
221-0037 221-0039 221-0041 221-0044 221-0040 221-0038 221-0042 221-0045 221-0168
Funnels, industrial, Azlon®
Funis Funis de filtração Funis de filtração
HDPE, translucent.
  • Good chemical and temperature resistance
  • Robust, rigid design
  • Feature straight sided upper position to prevent spillage
With smooth internal walls. Multipurpose funnel for a wide variety of uses. Suspension eyelet for easy storage.