Print…
Vent filters, HEPA-CAP, Whatman™
Fornecedor: Whatman products (Cytiva)
6702-3600 6702-7500 2609T 6702-9500 2709T
516-1104EA 69.75 EUR
516-1104 516-1105 517-0008 517-0012 517-0011
Vent filters, HEPA-CAP, Whatman™
Filtros Filtros de Ventilação
High-retention and high flow-rate glass fiber filters for research and industrial use to remove particulates from air and gases.
  • Typical retention efficiency: 99.97% of all particles ≥ 0.3 µm in air
  • Depth filter design: Allows for high loading capacity and bi-directional flow
  • Repeatedly autoclavable: At 121 °C for 20 minutes (132 °C max.)
Whatman HEPA-CAP Vent Filters are used across scientific, research, and industrial environments in a variety of air and gas filtration applications that require high particle retention and the ability to capture dirtMais informação do produto
Encomendar
ESPECIFICAÇÕES
Environmentally Preferable
Ø 36 mm 75 mm 36 mm 150 mm 75 mm
Tipo de caixa cápsula cápsula Hepa-Cap™ 36 capsule Hepa-Cap™ 150 capsule Hepa-Cap™ 75 capsule
Débito de ar 225 L/min (0,35 bar) 700 L/min (0,35 bar) 225 L/min (0,35 bar) 225 L/min (0,35 bar) 225 L/min (0,35 bar)
Pressão máx. 4,1 bar 4,1 bar 4,1 bar 4,1 bar 4,1 bar
Entrada/saída 1/4 to 3/8" stepped barb 3/8 to 1/2" stepped barb 3/8" FNPT 3/8" FNPT 3/8" FNPT