Print…
Tubing connector, straight
Fornecedor: Brand
152400 152715 152705 152710
229-3176EA 40.3 EUR
229-3176 229-3173 229-3171 229-3172
Tubing connector, straight
Conexões e Conectores
PP, transparent, with tapered ends.
  • Good mechanical and chemical resistance
  • Very good thermal resistance
For tubes with varying diameters.