Print…
Columns, TSK-Gel® Amide-80
Fornecedor: TOSOH Bioscience
0013071 0019021 0019697 0019696 0021865 0014459 0014460 0019694 0019695 0023455 0023454 0023456 0023457 0023458 0023459 0023460
554-1412EA 1220 EUR
554-1412 554-1650 554-1415 554-1414 554-1601 554-1622 554-1623 554-1651 554-1652 TOYO0023455 TOYO0023454 554-5942 TOYO0023457 TOYO0023458 TOYO0023459 TOYO0023460
Columns, TSK-Gel® Amide-80
Cromatografia, Colunas
Stainless steel columns with non-ionic carbamoyl [-C(NH₂)O] groups on a silica gel basis suitable for the separation of hydrophilic and polar compounds.
Max. pressure: 150 kg/cm²
Organic solvents: 0–100%
pH: 2 to 7,5
Salt concentration: 100 mM
Temperature stability: 4 - 80 °C