Print…
Pinças para tubos, PVDF, LaboPlast®
Fornecedor: BURKLE
8619-0104 8619-0102 8619-0202 8619-0206 8619-0154 8619-0155 8619-0105 8619-0106 8619-0152 8619-0156 8619-0205 8619-0204
229-0608EA 29.6 EUR
229-0608 229-0607 229-0613 229-0616 229-0618 229-0611 229-0609 229-0610 229-0617 229-0612 229-0615 229-0614
Pinças para tubos, PVDF, LaboPlast®
Pinças /Clamps Pinças para tubos
PVDF, aluminium and galvanised steel hook. The micro adjustment screw enables the flow of liquids to be controlled delicately and smoothly or stopped completely. The clamp can also be mounted on existing tube connections. The clamp's closure can be opened in any position, without screwing on and off.
  • Heat resistant up to +140 °C
  • Various sizes available for tubing from 1 to 20 mm Ext.Ø
  • Available in four colours