Print…
Clamps
Fornecedor: USBECK Laborgeräte
2510 2520 2530
241-7317EA 10.7 EUR
241-7317 241-0252 241-0253
Clamps
Pinças /Clamps Pinças de uso geral
Zinc die-cast, powder-coated.
  • Cork lined jaws
  • With rod (Ø 10 mm)
  • One jaw in V-shape
Length: 250 mm