Print…
Burettes, straight stopcock, Azlon®
Fornecedor: SciLabware
BDH112T BDH118T
612-0348EA 113 EUR
612-0348 612-0349
Burettes, straight stopcock, Azlon®
Buretas
Acrylic, with PMP/PE stopcocks, class B.
  • Graduations are screen printed to ISO 385 class B accuracy
  • Unaffected by most dilute aqueous solutions (except ammonia and hydrofluoric acid) but unsuitable for use with ethanol or organic solvents
  • Suitable for both left and right handed use
Acrylic construction for strength and transparency. Suitable for use in education, research or industrial laboratories, especially where breakages often occur.
Fornecimento: Supplied with a technical data sheet.