Print…
Frascos, de boca estreita, quadrados, com tampa roscada, Nalgene®
Fornecedor: Thermo Fisher Scientific
2016-0030 2016-0060 2016-0125 2016-0250 2016-1000 2018-0030 2018-0060 2018-0125 2018-0250 2018-0500 2018-1000 2016-0500
215-7621EA 19.8 EUR
215-7621 215-7622 215-7623 215-7624 215-7626 215-7641 215-7642 215-7643 215-7644 215-7645 215-7646 215-7625
Frascos, de boca estreita, quadrados, com tampa roscada, Nalgene®
Frascos Frascos de Plástico
De HDPE ou de PPCO, translúcidos, com tampa roscada de PP.
  • HDPE: Good chemical resistance/PPCO: Very good chemical resistance
  • HDPE: Heat resistant up to 120 °C/PPCO: Heat resistant up to 135 °C, autoclavable
  • Leakproof
Ideal for storing and shipping liquids. Graduated.