Print…
Beakers, low form
Fornecedor: NICKEL ELECTRO
402 404 407 410 412 415
213-0226EA 73.3 EUR
213-0226 213-0227 213-0228 213-0229 213-0230 NICK415
Beakers, low form
Copos
Aluminium, with spout.
  • Good chemical resistance
  • Excellent resistance to heat
  • Smooth surfaces for fast, easy cleaning
Ideal for corrosive materials. Robust and durable.