Print…
Beakers, low form, Azlon®
Fornecedor: SciLabware
BDA230P BDA212P BDA218P BDA224P BDA236P BDA240P BDA242P
213-0064EA 97.7 EUR
213-0064 213-0058 213-0060 213-0062 213-0067 213-0070 213-0072
Beakers, low form, Azlon®
Copos
PP, translucent.
  • Good chemical resistance
  • Resistant to heat up to 135 °C
  • Smooth surfaces for quick, easy cleaning
Ideal for general laboratory use. With spout and exceptionally easy to read blue graduation. Straight sided.
ISO 7056
Certificações: ISO 7056.