Print…
Beakers, tall form
Fornecedor: SciLabware
1015/06D 1015/02M 1015/04D 1015/14D 1015/18D 1015/22D 1015/10D 1015/26D
213-0033EA 69.8 EUR
213-0033 213-0031 213-0032 213-0035 213-0036 213-0037 213-0034 213-0415
Beakers, tall form
Copos
PYREX®, borosilicate glass, with spout.
  • Very good chemical resistance
  • High temperature resistance
  • Autoclavable
Volume graduations and labelling area.
Certificações: DIN 12331, ISO 3819