Print…
EasySep™ Human Whole Blood CD34 Positive Selection Kit II
Fornecedor: STEMCELL TECHNOLOGIES
17879 17879RF
STMC17879EA 1410 EUR
STMC17879 STMC17879RF
EasySep™ Human Whole Blood CD34 Positive Selection Kit II
Meios de separação celular
Immunomagnetic positive selection of human CD34+ cells from whole blood or buffy coat.
  • Fast and easy-to-use
  • No columns required
The EasySep™ Human Whole Blood CD34 Positive Selection Kit II is designed to isolate CD34+ cells from fresh whole blood or freshly prepared buffy coat by positive selection. Desired cells are targeted with Tetrameric Antibody Complexes recognising CD34 and dextran-coated magnetic particles. The cocktail also contains an antibody to human Fc receptor to minimize nonspecific binding. labelled cells are separated using an EasySep™ magnet without the use of columns. Cells of interest remain in the tube while unwanted cells are poured off. The CD34 antigen is expressed on hematopoietic stem and progenitor cells.
This product can be used instead of the EasySep™ Human Whole Blood CD34 Positive Selection Kit should be used.
Atenção: Product Restrictions: (i) Customer shall not make products or any portion of them, in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier's mark of origin without express written permission of Supplier; and (ii) Customer is familiar with Supplier's restrictions on the use of the Products and shall use the Products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').