Print…
3-[(3-Colamidopropil)dimetilamónio]-1-propano sulfato (CHAPS), grade proteomic
Fornecedor: VWR Chemicals
Atenção
Sinônimos: CHAPS
M127-10GEA 287 EUR
M127-10G M127-5G
3-[(3-Colamidopropil)dimetilamónio]-1-propano sulfato (CHAPS), grade proteomic
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Detergente Zwitteriônico.
  • High purity and protease-free
  • Suitable for most biological applications
  • Both positively and negatively charged
These mild detergents efficiently solubilise membrane proteins without affecting the native state or activity of the proteins. They are more efficient at preventing non-specific protein aggregation than ionic detergents and are easily removed from solutions by dialysis. Zwitterionic detergents are frequently used in electrophoresis and isoelectric focusing, as well as ion-exchange chromatography.
Fórmula: C₃₂H₅₈N₂O₇S
Ponto de Fusão: 116 °C
Temperatura de Armazenamento: Frigorífico
Número MDL : MFCD00081083
Número CAS: 75621-03-3
Merck Index: 12,02085
Encomendar
Conductivity (10%, Water)
DNase
pH (10%, Water) @25 °C
Protease
Purity (TLC)
Residue on Ignition
RNase
Solubility (20 %, Water)