Print…

Z-Mirin

Fornecedor: TCI

M3303-100MG M3303-25MG
TCIAM3303-100MGEA 190 EUR
TCIAM3303-100MG TCIAM3303-25MG
Z-Mirin
Z-Mirin
Fórmula: C₁₀H₈N₂O₂S
Número MDL : MFCD05885480
Número CAS: 1198097-97-0

Encomendar


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR