Print…
Anti-IgG Goat Polyclonal Antibody (mFluor™ UV 375)
Fornecedor: AAT BIOQUEST
49450 49451
AATB49450EA 266 EUR
AATB49450 AATB49451
Anti-IgG Goat Polyclonal Antibody (mFluor™ UV 375)
Anticorpos
mFluor™ UV375 goat anti-mouse conjugates are secondary antibodies designed for optimal performance in immunoassay applications, including flow cytometry, immunofluorescence, and confocal microscopy.
  • Purified and quality-tested for consistent results
  • Suitable for multiple applications
  • Purified through affinity chromatography
mFluor™ UV375 is optimally excited by the UV laser and emits maximally at 387 nm.
Atenção: Research use only.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: mFluor™ UV 375
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Mouse