CM-DiI, CytoTrace™
Fornecedor: AAT BIOQUEST
1,1'-Dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine iodide (CM-DiI) is a lipophilic carbocyanine dye that is commonly used for labeling cell membranes in various biological and neuroscientific research applications.
- Stable for 12 months
Quality tested in-house
Atenção: For Research Use Only