Print…

Anti-Estrogen Inducible Protein pS2 Mouse Monoclonal Antibody [clone: ABT-PS2]

Fornecedor: ANTIBODIES.COM
A100699-100 A100699-50
ANTIA100699-100EA 475 EUR
ANTIA100699-100 ANTIA100699-50
Anti-Estrogen Inducible Protein pS2 Mouse Monoclonal Antibody [clone: ABT-PS2]
Anticorpos
Mouse monoclonal (ABT-PS2) antibody to pS2 for IHC with samples derived from human.

  • Validated Applications: IHC

Type: Primary
Antigen: Estrogen Inducible Protein pS2
Clonality: Monoclonal
Clone: ABT-PS2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR