Print…
Anti-CD35 Mouse Monoclonal Antibody [clone: ABT-CD35]
Fornecedor: ANTIBODIES.COM
A100031-100 A100031-50
ANTIA100031-100EA 475 EUR
ANTIA100031-100 ANTIA100031-50
Anti-CD35 Mouse Monoclonal Antibody [clone: ABT-CD35]
Anticorpos
Mouse monoclonal (ABT-CD35) antibody to CD35 for IHC with samples derived from human.
  • Validated Applications: IHC
Type: Primary
Antigen: CD35
Clonality: Monoclonal
Clone: ABT-CD35
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human