Print…

Anti-CD7 Mouse Monoclonal Antibody [clone: ABT-CD7]

Fornecedor: ANTIBODIES.COM
A100702-100 A100702-50
ANTIA100702-100EA 475 EUR
ANTIA100702-100 ANTIA100702-50
Anti-CD7 Mouse Monoclonal Antibody [clone: ABT-CD7]
Anticorpos
Mouse monoclonal (ABT-CD7) antibody to CD7 for IHC with samples derived from human.

  • Validated Applications: IHC

Type: Primary
Antigen: CD7
Clonality: Monoclonal
Clone: ABT-CD7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR