Print…
Anti-p63 Protein + HMW Cytokeratin Mouse Monoclonal Antibody [clone: ABT-P40]
Fornecedor: ANTIBODIES.COM
A100700-100 A100700-50
ANTIA100700-100EA 475 EUR
ANTIA100700-100 ANTIA100700-50
Anti-p63 Protein + HMW Cytokeratin Mouse Monoclonal Antibody [clone: ABT-P40]
Anticorpos
Mouse monoclonal (ABT-P40) antibody to p40 for IHC with samples derived from human.
  • Validated Applications: IHC
Type: Primary
Antigen: p63 Protein + HMW Cytokeratin
Clonality: Monoclonal
Clone: ABT-P40
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human