Print…
Anti-CD34 Mouse Monoclonal Antibody [clone: ABT-CD34]
Fornecedor: ANTIBODIES.COM
A100032-100 A100032-50
ANTIA100032-100EA 475 EUR
ANTIA100032-100 ANTIA100032-50
Anti-CD34 Mouse Monoclonal Antibody [clone: ABT-CD34]
Anticorpos
Mouse monoclonal (ABT-CD34) antibody to CD34 for IHC with samples derived from human.
  • Validated Applications: IHC
Type: Primary
Antigen: CD34
Clonality: Monoclonal
Clone: ABT-CD34
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human