Print…
Anti-Cytokeratin 10 + 13 Mouse Monoclonal Antibody [clone: ABT-CK10]
Fornecedor: ANTIBODIES.COM
A96673-100 A96673-50
ANTIA96673-100EA 475 EUR
ANTIA96673-100 ANTIA96673-50
Anti-Cytokeratin 10 + 13 Mouse Monoclonal Antibody [clone: ABT-CK10]
Anticorpos
Mouse monoclonal (ABT-CK10) antibody to Cytokeratin 10 for IHC with samples derived from Human.
  • Validated Applications: IHC
Type: Primary
Antigen: Cytokeratin 10 + 13
Clonality: Monoclonal
Clone: ABT-CK10
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human