Print…

Anti-p63 Protein + HMW Cytokeratin Mouse Monoclonal Antibody [clone: ABT-CK16]

Fornecedor: ANTIBODIES.COM
A96672-100 A96672-50
ANTIA96672-100EA 475 EUR
ANTIA96672-100 ANTIA96672-50
Anti-p63 Protein + HMW Cytokeratin Mouse Monoclonal Antibody [clone: ABT-CK16]
Anticorpos
Mouse monoclonal (ABT-CK16) antibody to Cytokeratin 16 for IHC with samples derived from Human.

  • Validated Applications: IHC

Type: Primary
Antigen: p63 Protein + HMW Cytokeratin
Clonality: Monoclonal
Clone: ABT-CK16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR