Print…

Anti-p63 Protein + HMW Cytokeratin Mouse Monoclonal Antibody [clone: ABT-CK13]

Fornecedor: ANTIBODIES.COM
A95697-100 A95697-50
ANTIA95697-100EA 475 EUR
ANTIA95697-100 ANTIA95697-50
Anti-p63 Protein + HMW Cytokeratin Mouse Monoclonal Antibody [clone: ABT-CK13]
Anticorpos
Mouse monoclonal (ABT-CK13) antibody to Cytokeratin 13 for IHC and WB with samples derived from Human.

  • Validated Applications: IHC, WB

Type: Primary
Antigen: p63 Protein + HMW Cytokeratin
Clonality: Monoclonal
Clone: ABT-CK13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR