Print…

Anti-EpCAM Mouse Monoclonal Antibody [clone: ABT-EPCAM]

Fornecedor: ANTIBODIES.COM
A99689-100 A99689-50
ANTIA99689-100EA 475 EUR
ANTIA99689-100 ANTIA99689-50
Anti-EpCAM Mouse Monoclonal Antibody [clone: ABT-EPCAM]
Anticorpos
Mouse monoclonal (ABT-EPCAM) antibody to EpCAM for IHC with samples derived from human.

  • Validated applications: IHC

Type: Primary
Antigen: EpCAM
Clonality: Monoclonal
Clone: ABT-EPCAM
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR