Print…

Anti-HDAC6 Mouse Monoclonal Antibody [clone: ABT-HDAC6]

Fornecedor: ANTIBODIES.COM
A99688-100 A99688-50
ANTIA99688-100EA 475 EUR
ANTIA99688-100 ANTIA99688-50
Anti-HDAC6 Mouse Monoclonal Antibody [clone: ABT-HDAC6]
Anticorpos
Mouse monoclonal (ABT-HDAC6) antibody to HDAC6 for IHC with samples derived from human.

  • Validated applications: IHC

Type: Primary
Antigen: HDAC6
Clonality: Monoclonal
Clone: ABT-HDAC6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR