Print…

Anti-S100P Mouse Monoclonal Antibody [clone: ABT-S100P]

Fornecedor: ANTIBODIES.COM
A99686-100 A99686-50
ANTIA99686-100EA 475 EUR
ANTIA99686-100 ANTIA99686-50
Anti-S100P Mouse Monoclonal Antibody [clone: ABT-S100P]
Anticorpos
Mouse monoclonal (ABT-S100P) antibody to S100P for IHC with samples derived from human.

  • Validated applications: IHC

Type: Primary
Antigen: S100P
Clonality: Monoclonal
Clone: ABT-S100P
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR