Print…

Anti-Proinsulin Mouse Monoclonal Antibody [clone: ABT-INS]

Fornecedor: ANTIBODIES.COM
A99499-100 A99499-50
ANTIA99499-100EA 475 EUR
ANTIA99499-100 ANTIA99499-50
Anti-Proinsulin Mouse Monoclonal Antibody [clone: ABT-INS]
Anticorpos
Mouse monoclonal (ABT-INS) antibody to Insulin for IHC with samples derived from human.

  • Validated applications: IHC

Type: Primary
Antigen: Proinsulin
Clonality: Monoclonal
Clone: ABT-INS
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR