Print…
Anti-CD44v6 Mouse Monoclonal Antibody [clone: ABT-CD44]
Fornecedor: ANTIBODIES.COM
A99231-100 A99231-50
ANTIA99231-100EA 475 EUR
ANTIA99231-100 ANTIA99231-50
Anti-CD44v6 Mouse Monoclonal Antibody [clone: ABT-CD44]
Anticorpos
Mouse monoclonal (ABT-CD44) antibody to CD44 for IHC and WB with samples derived from human.
  • Validated applications: IHC, WB
Type: Primary
Antigen: CD44v6
Clonality: Monoclonal
Clone: ABT-CD44
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human