Print…

Anti-S100A4 Mouse Monoclonal Antibody [clone: ABT-S100A4]

Fornecedor: ANTIBODIES.COM
A98938-100 A98938-50
ANTIA98938-100EA 475 EUR
ANTIA98938-100 ANTIA98938-50
Anti-S100A4 Mouse Monoclonal Antibody [clone: ABT-S100A4]
Anticorpos
Mouse monoclonal (ABT-S100A4) antibody to S100A4 for IHC with samples derived from human.

  • Validated applications: IHC

Type: Primary
Antigen: S100A4
Clonality: Monoclonal
Clone: ABT-S100A4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR