Print…
Anti-Myogenin Mouse Monoclonal Antibody [clone: ABT-MYOG]
Fornecedor: ANTIBODIES.COM
A98912-100 A98912-50
ANTIA98912-100EA 475 EUR
ANTIA98912-100 ANTIA98912-50
Anti-Myogenin Mouse Monoclonal Antibody [clone: ABT-MYOG]
Anticorpos
Mouse monoclonal (ABT-MYOG) antibody to Myogenin for IHC with samples derived from human.
  • Validated applications: IHC
Type: Primary
Antigen: Myogenin
Clonality: Monoclonal
Clone: ABT-MYOG
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human