Print…

Anti-Actin Mouse Monoclonal Antibody [clone: ABT-ACTN]

Fornecedor: ANTIBODIES.COM
A98880-100 A98880-50
ANTIA98880-100EA 475 EUR
ANTIA98880-100 ANTIA98880-50
Anti-Actin Mouse Monoclonal Antibody [clone: ABT-ACTN]
Anticorpos
Mouse monoclonal (ABT-ACTN) antibody to Actin for IHC and WB with samples derived from human.

  • Validated applications: IHC, WB

Type: Primary
Antigen:
Clonality: Monoclonal
Clone: ABT-ACTN
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR