Print…
Anti-Villin Mouse Monoclonal Antibody [clone: ABT-VIL]
Fornecedor: ANTIBODIES.COM
A98836-100 A98836-50
ANTIA98836-100EA 475 EUR
ANTIA98836-100 ANTIA98836-50
Anti-Villin Mouse Monoclonal Antibody [clone: ABT-VIL]
Anticorpos
Mouse monoclonal (ABT-VIL) antibody to Villin for IHC and WB with samples derived from human.
  • Validated applications: IHC, WB
Type: Primary
Antigen: Villin
Clonality: Monoclonal
Clone: ABT-VIL
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human