Print…

Anti-Vimentin Mouse Monoclonal Antibody [clone: ABT-VIM]

Fornecedor: ANTIBODIES.COM
A98367-100 A98367-50
ANTIA98367-100EA 475 EUR
ANTIA98367-100 ANTIA98367-50
Anti-Vimentin Mouse Monoclonal Antibody [clone: ABT-VIM]
Anticorpos
Mouse monoclonal (ABT-VIM) antibody to Vimentin for IHC and WB with samples derived from human.

  • Validated applications: IHC, WB

Type: Primary
Antigen: Vimentin
Clonality: Monoclonal
Clone: ABT-VIM
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR