Print…

Anti-Thyroglobulin Mouse Monoclonal Antibody [clone: ABT-TG]

Fornecedor: ANTIBODIES.COM
A98339-100 A98339-50
ANTIA98339-100EA 475 EUR
ANTIA98339-100 ANTIA98339-50
Anti-Thyroglobulin Mouse Monoclonal Antibody [clone: ABT-TG]
Anticorpos
Mouse monoclonal (ABT-TG) antibody to Thyroglobulin for IHC with samples derived from human.

  • Validated applications: IHC

Type: Primary
Antigen: Thyroglobulin
Clonality: Monoclonal
Clone: ABT-TG
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR