Print…
Anti-UCHL1+3 Mouse Monoclonal Antibody [clone: ABT-PGP9.5]
Fornecedor: ANTIBODIES.COM
A97080-100 A97080-50
ANTIA97080-100EA 475 EUR
ANTIA97080-100 ANTIA97080-50
Anti-UCHL1+3 Mouse Monoclonal Antibody [clone: ABT-PGP9.5]
Anticorpos
Mouse monoclonal (ABT-PGP9.5) antibody to PGP 9.5 for IHC and WB with samples derived from human.
  • Validated applications: IHC, WB
Type: Primary
Antigen: UCHL1+3
Clonality: Monoclonal
Clone: ABT-PGP9.5
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human