Print…
Anti-p63 Protein + HMW Cytokeratin Mouse Monoclonal Antibody [clone: ABT-P63]
Fornecedor: ANTIBODIES.COM
A99729-100 A99729-50
ANTIA99729-100EA 475 EUR
ANTIA99729-100 ANTIA99729-50
Anti-p63 Protein + HMW Cytokeratin Mouse Monoclonal Antibody [clone: ABT-P63]
Anticorpos
Mouse monoclonal (ABT-P63) antibody to p63 for IHC and WB with samples derived from Human.
  • Validated applications: IHC, WB
Type: Primary
Antigen: p63 Protein + HMW Cytokeratin
Clonality: Monoclonal
Clone: ABT-P63
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human