Print…
Anti-IgG Goat Polyclonal Antibody (iFluor® 450)
Fornecedor: AAT BIOQUEST
50052 50053
AATB50052EA 120 EUR
AATB50052 AATB50053
Anti-IgG Goat Polyclonal Antibody (iFluor® 450)
Anticorpos
Anti-human secondary antibodies have well-characterised specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target.
  • Affinity purified
  • Easily reconstituted
Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence).
Atenção: For research use only.
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: iFluor® 450
Epitope:
Host: Goat
Isotype: IgG (H, L)
Reactivity: Human