Print…

Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)

Fornecedor: AAT BIOQUEST
100331U0 100331U1 100331U2
AATB100331U0EA 319 EUR
AATB100331U0 AATB100331U1 AATB100331U2
Anti-CD3e Mouse Monoclonal Antibody [clone: SK7] (PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®)
Anticorpos
The SK7 monoclonal antibody reacts with human CD3e, a 20 kD member of the Ig superfamily often found on the surface of t cells, tregs and nkt cells.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Atenção: For research use only.

Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: SK7
Conjugation: PerCP (Peridinin-Chlorophyll Protein Complex)/Cy5.5®
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR