Print…
Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (PE/Texas Red®)
Fornecedor: AAT BIOQUEST
101411S0 101411S1 101411S2
AATB101411S0EA 319 EUR
AATB101411S0 AATB101411S1 AATB101411S2
Anti-CD14 Mouse Monoclonal Antibody [clone: 61D3] (PE/Texas Red®)
Anticorpos
61D3 is an anti-human monoclonal antibody that targets the CD14 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Atenção: For research use only.
Type: Primary
Antigen: CD14 antigen
Clonality: Monoclonal
Clone: 61D3
Conjugation: PE/Texas Red®
Epitope:
Host: Mouse
Isotype:
Reactivity: Human