Print…
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (PE/Cy7)
Fornecedor: AAT BIOQUEST
100511N0 100511N1 100511N2
AATB100511N0EA 319 EUR
AATB100511N0 AATB100511N1 AATB100511N2
Anti-CD5 Mouse Monoclonal Antibody [clone: L17F12] (PE/Cy7)
Anticorpos
L17F12 is an anti-human monoclonal antibody that is specific for the CD5 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Atenção: For research use only.
Type: Primary
Antigen: CD5 antigen
Clonality: Monoclonal
Clone: L17F12
Conjugation: PE/Cy7
Epitope:
Host: Mouse
Isotype:
Reactivity: Human