Print…

Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (mFluor™ Violet 450)

Fornecedor: AAT BIOQUEST
104410Z0 104410Z1
AATB104410Z0EA 371 EUR
AATB104410Z0 AATB104410Z1
Anti-CD44 Rat Monoclonal Antibody [clone: IM7] (mFluor™ Violet 450)
Anticorpos
IM7 is an anti-human/mouse monoclonal antibody that targets the CD44 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Atenção: For research use only.

Type: Primary
Antigen: CD44 antigen
Clonality: Monoclonal
Clone: IM7
Conjugation: mFluor™ Violet 450
Epitope:
Host: Rat
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR