Print…

Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (mFluor™ Red 780)

Fornecedor: AAT BIOQUEST
101920W0 101920W1
AATB101920W0EA 371 EUR
AATB101920W0 AATB101920W1
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (mFluor™ Red 780)
Anticorpos
HIB19 is an anti-human monoclonal antibody that recognises the CD19 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Atenção: For research use only.

Type: Primary
Antigen: CD19 antigen
Clonality: Monoclonal
Clone: HIB19
Conjugation: mFluor™ Red 780
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR