Print…
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (mFluor™ Blue 570)
Fornecedor: AAT BIOQUEST
101920T0 101920T1
AATB101920T0EA 371 EUR
AATB101920T0 AATB101920T1
Anti-CD19 Mouse Monoclonal Antibody [clone: HIB19] (mFluor™ Blue 570)
Anticorpos
HIB19 is an anti-human monoclonal antibody that recognises the CD19 antigen.
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Atenção: For research use only.
Type: Primary
Antigen: CD19 antigen
Clonality: Monoclonal
Clone: HIB19
Conjugation: mFluor™ Blue 570
Epitope:
Host: Mouse
Isotype:
Reactivity: Human