Print…

Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (iFluor® 405)

Fornecedor: AAT BIOQUEST
10340020 10340021
AATB10340020EA 371 EUR
AATB10340020 AATB10340021
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (iFluor® 405)
Anticorpos
4H11 is an anti-human monoclonal antibody that recognises the CD34 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Atenção: For research use only.

Type: Primary
Antigen: CD34 antigen
Clonality: Monoclonal
Clone: 4H11
Conjugation: iFluor® 405
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR