Print…

Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (APC/Cy7)

Fornecedor: AAT BIOQUEST
103801D0 103801D1 103801D2
AATB103801D0EA 319 EUR
AATB103801D0 AATB103801D1 AATB103801D2
Anti-CD38 Mouse Monoclonal Antibody [clone: HIT2] (APC/Cy7)
Anticorpos
HIT2 is an anti-human monoclonal antibody that targets the CD38 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Atenção: For research use only.

Type: Primary
Antigen: CD38 antigen
Clonality: Monoclonal
Clone: HIT2
Conjugation: APC/Cy7
Epitope:
Host: Mouse
Isotype: Mouse IgG1
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR