Print…
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (PE/iFluor® 594)
Fornecedor: AAT BIOQUEST
100321Y0 100321Y1 100321Y2
AATB100321Y0EA 319 EUR
AATB100321Y0 AATB100321Y1 AATB100321Y2
Anti-CD3e Mouse Monoclonal Antibody [clone: UCHT1] (PE/iFluor® 594)
Anticorpos
The UCHT1 monoclonal antibody binds to human CD3e, a 20 kD member of the Ig superfamily commonly found on the surface of tregs, thymocytes, nkt cells and thymocytes (differentiation dependents).
  • Particularly useful for flow cytometry
This antibody was purified through affinity chromatography.
Atenção: For research use only.
Type: Primary
Antigen: CD3e
Clonality: Monoclonal
Clone: UCHT1
Conjugation: PE/iFluor® 594
Epitope:
Host: Mouse
Isotype:
Reactivity: Human