Print…

Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® A7)

Fornecedor: AAT BIOQUEST
116100S0 116100S1
AATB116100S0EA 371 EUR
AATB116100S0 AATB116100S1
Anti-CD161 Mouse Monoclonal Antibody [clone: HP-3G10] (iFluor® A7)
Anticorpos
HP-3G10 is an anti-human monoclonal antibody that recognises the CD161 antigen.

  • Particularly useful for flow cytometry

This antibody was purified through affinity chromatography.

Atenção: For research use only.

Type: Primary
Antigen: CD161
Clonality: Monoclonal
Clone: HP-3G10
Conjugation: iFluor® A7
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR